2-Hydroxypyrimidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Pyrimidinol Hydrochloride2(1H)-pyrimidinone monohydrochloridePyrimidin-2-ol hydrochloride

074890-100G
102538-716EA 184.62 USD
102538-716
2-Hydroxypyrimidine hydrochloride ≥95%
2-Hydroxypyrimidine hydrochloride

Hazard: Irritant

Formula: C₄H₄N₂O
MW: 132.55 g/mol
MDL Number: MFCD00012781
CAS Number: 38353-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR