(R)-(+)-N-Benzyl-α-methylbenzylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-(+)-N-Benzyl-1-phenylethylamine
, (R)-N-Benzyl-1-phenylethanamine
074889-100G
102538-712EA
156.92
USD
102538-712
(R)-(+)-N-Benzyl-α-methylbenzylamine ≥95%
(R)-(+)-N-Benzyl-α-methylbenzylamine
Hazard: Irritant/Air Sensitive
Formula:
C₁₅H₁₇N MW: 211.31 g/mol Boiling Pt: 171 °C (15 mmHg) Density: 1.01 |
MDL Number:
MFCD00015010 CAS Number: 38235-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...