(R)-(-)-Ethyl-3-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-Ethyl piperidine-3-carboxylate

074879-5G
102538-660EA 78.46 USD
102538-660
(R)-(-)-Ethyl-3-piperidinecarboxylate ≥95%
(R)-(-)-Ethyl 3-piperidinecarboxylate

Hazard: Irritant

Formula: C₈H₁₅NO₂
MW: 157.2102 g/mol
MDL Number: MFCD03093637
CAS Number: 25137-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR