3,5-Dichloropyridine ≥95%

Supplier: Matrix Scientific

074877-100G
102538-652EA 200 USD
102538-652
3,5-Dichloropyridine ≥95%
3,5-Dichloropyridine

Hazard: Toxic

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 178 °C
Melting Pt: 63…67 °C
Density: 1.39
MDL Number: MFCD00006376
CAS Number: 2457-47-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR