N,O-Dimethylhydroxylamine hydrochloride ≥95%

Supplier: Matrix Scientific

074874-100G
102538-640EA 52.31 USD
102538-640
N,O-Dimethylhydroxylamine hydrochloride ≥95%
N,O-Dimethylhydroxylamine hydrochloride

Hazard: Irritant

Formula: C₂H₈ClNO
MW: 97.55 g/mol
MDL Number: MFCD00012485
CAS Number: 6638-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR