2-Piperidineethanol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(2-Hydroxyethyl)piperidine
074863-100G
074863-10G
102538-588EA
201.54
USD
102538-588
102538-586
2-Piperidineethanol ≥95%
2-Piperidineethanol
Hazard: Irritant
Formula:
C₇H₁₅NO MW: 129.20 g/mol |
MDL Number:
MFCD00005989 CAS Number: 1484-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...