1,3-Dichloro-5,5-dimethylhydantoin ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Dichloro-5,5-dimethylhydantoine1,3-Dichloro-5,5-dimethylimidazolidine-2,4-dione

074847-100G
102538-518EA 92.31 USD
102538-518
1,3-Dichloro-5,5-dimethylhydantoin ≥95%
1,3-Dichloro-5,5-dimethylhydantoin

Hazard: Irritant

Formula: C₅H₆Cl₂N₂O₂
MW: 197.02 g/mol
MDL Number: MFCD00003190
CAS Number: 118-52-5
UN: 1479
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR