(2-Hydroxyethyl)hydrazine ≥95%

Supplier: Matrix Scientific

Synonyms: β-Hydroxyethylhydrazine

074844-100G
102538-506EA 172.5 USD
102538-506
(2-Hydroxyethyl)hydrazine ≥95%
(2-Hydroxyethyl)hydrazine

Hazard: Toxic

Formula: C₂H₈N₂O
MW: 76.1 g/mol
Boiling Pt: 218…220 °C
Density: 1.121
MDL Number: MFCD00007623
CAS Number: 109-84-2
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR