3-Aminobenzoic acid ≥95%

Supplier: Matrix Scientific

074830-100G 074830-500G
102538-448EA 43.08 USD
102538-448 102833-722
3-Aminobenzoic acid ≥95%
3-Aminobenzoic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00007795
CAS Number: 99-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR