(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥95%

Supplier: Matrix Scientific

Synonyms: PyBOP1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)

074823-500G 074823-100G
102551-916EA 361.54 USD
102551-916 102538-418
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥95%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Hazard: Irritant/Store in Freezer

Formula: C₁₈H₂₈F₆N₆OP₂
MW: 520.4 g/mol
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR