Diphenylmethylene-glycine benzyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
N-(Diphenylmethylene) glycine benzyl ester
074790-5G
102538-236EA
107.69
USD
102538-236
Diphenylmethylene-glycine benzyl ester ≥95%
Diphenylmethylene-glycine benzyl ester
Hazard: Irritant
Formula:
C₂₂H₁₉NO₂ MW: 329.3900 g/mol |
MDL Number:
MFCD08062206 CAS Number: 81477-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...