4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

074787-1G 074787-5G
102538-220EA 923.08 USD
102538-220 102538-222
4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester ≥95%
4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₈H₂₉BN₂O₂
MW: 316.2461 g/mol
MDL Number: MFCD13190738
CAS Number: 656257-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR