N,N-Dimethylthioformamide ≥95%

Supplier: Matrix Scientific

074779-5G
102538-178EA 69.23 USD
102538-178
N,N-Dimethylthioformamide ≥95%
N,N-Dimethylthioformamide

Hazard: Irritant

Formula: C₃H₇NS
Boiling Pt: 58…60 °C (1 mmHg)
MDL Number: MFCD00004943
CAS Number: 758-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR