2-(4-Iodophenyl)ethanol ≥95%

Supplier: Matrix Scientific

074776-1G 074776-5G
102538-164EA 233.85 USD
102538-164 102538-166
2-(4-Iodophenyl)ethanol ≥95%
2-(4-Iodophenyl)ethanol

Hazard: Irritant

Formula: C₈H₉IO
MW: 248.0600 g/mol
MDL Number: MFCD00173705
CAS Number: 52914-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR