4-Morpholinocyclohexanone ≥95%

Supplier: Matrix Scientific

074771-1G 074771-250MG
102538-142EA 684.62 USD
102538-142 102538-144
4-Morpholinocyclohexanone ≥95%
4-Morpholinocyclohexanone

Hazard: Irritant

Formula: C₁₀H₁₇NO₂
MW: 183.2500 g/mol
MDL Number: MFCD17013167
CAS Number: 139025-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR