Methyl-5-formylthiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

074763-5G 074763-1G
102538-100EA 176.92 USD
102538-100 102538-102
Methyl-5-formylthiophene-3-carboxylate ≥95%
Methyl-5-formylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₇H₆O₃S
MW: 170.19
MDL Number: MFCD01859945
CAS Number: 67808-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR