1-N-Cbz-4-aminopiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-N-Cbz-4-AminopiperidineBenzyl 4-Amino-1-piperidinecarboxylate4-Amino-1-carbobenzoxypiperidineBenzyl 4-aminopiperidine-1-carboxylate4-Amino-1-piperidinecarboxylic acid Benzyl ester4-Amino-1-Cbz-piperidine4-Amino-1-benzyloxycarbonylpiperidine

074761-5G
102538-090EA 230.77 USD
102538-090
1-N-Cbz-4-aminopiperidine ≥95%
1-N-Cbz-4-aminopiperidine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.30 g/mol
MDL Number: MFCD05863884
CAS Number: 120278-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR