Methyl-6-fluoronicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-fluoropyridine-3-carboxylate
, 6-Fluoronicotinic acid methyl ester
074746-5G
074746-1G
102538-010EA
343.08
USD
102538-010
102538-008
Methyl-6-fluoronicotinate ≥95%
Methyl 6-fluoronicotinate
Hazard: Irritant
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol |
MDL Number:
MFCD03095098 CAS Number: 1427-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...