4-Bromo-1H-pyrazole-3-carboxamide ≥95%

Supplier: Matrix Scientific

074742-250MG 074742-1G 074742-5G
102537-990EA 2061.54 USD
102537-990 102537-986 102537-988
4-Bromo-1H-pyrazole-3-carboxamide ≥95%
4-Bromo-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₄H₄BrN₃O
CAS Number: 932-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR