Methyl-4-bromofuran-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromofuran-2-carboxylate
074734-5G
074734-250MG
074734-1G
102537-940EA
498.46
USD
102537-940
102537-942
102537-944
Methyl-4-bromofuran-2-carboxylate ≥95%
Methyl-4-bromofuran-2-carboxylate
Hazard: Irritant
Formula:
C₆H₅BrO₃ MW: 205.0100 g/mol |
MDL Number:
MFCD00092559 CAS Number: 58235-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...