2-Chloro-3-(2-methoxyethylamino)naphthalene-1,4-dione ≥95%

Supplier: Matrix Scientific

074730-1G 074730-250MG
102537-920EA 684.62 USD
102537-920 102537-922
2-Chloro-3-(2-methoxyethylamino)naphthalene-1,4-dione ≥95%
2-Chloro-3-(2-methoxyethylamino)naphthalene-1,4-dione

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₃
MW: 265.6900 g/mol
MDL Number: MFCD02998090
CAS Number: 22272-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR