1-Cyclobutylethanol ≥95%

Supplier: Matrix Scientific

074718-250MG 074718-1G
102537-854EA 281.52 USD
102537-854 102537-856
1-Cyclobutylethanol ≥95%
1-Cyclobutylethanol

Hazard: Irritant

Formula: C₆H₁₂O
MW: 100.1589 g/mol
MDL Number: MFCD00061039
CAS Number: 7515-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR