Ethyl-6,8-dichlorooctanoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6,8-dichlorooctanoate

074717-10G 074717-5G 074717-1G
102537-850EA 278.46 USD
102537-850 102537-852 102537-848
Ethyl-6,8-dichlorooctanoate ≥95%
Ethyl-6,8-dichlorooctanoate

Hazard: Irritant

Formula: C₁₀H₁₈Cl₂O₂
MW: 241.1600 g/mol
MDL Number: MFCD01711127
CAS Number: 1070-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR