4-Chloro-3,5-difluorobenzonitrile ≥95%
Supplier: Matrix Scientific
074715-1G
074715-250MG
102537-840EA
684.62
USD
102537-840
102537-842
4-Chloro-3,5-difluorobenzonitrile ≥95%
4-Chloro-3,5-difluorobenzonitrile
Hazard: Irritant
Formula:
C₇H₂ClF₂N MW: 173.5500 g/mol |
MDL Number:
MFCD18415564 CAS Number: 144797-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...