4-Chloro-2,5-difluorobenzaldehyde ≥95%
Supplier: Matrix Scientific
074706-1G
074706-10G
074706-5G
102537-794EA
143.08
USD
102537-794
102537-798
102537-796
4-Chloro-2,5-difluorobenzaldehyde ≥95%
4-Chloro-2,5-difluorobenzaldehyde
Hazard: Irritant
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol |
MDL Number:
MFCD07782051 CAS Number: 879093-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...