4-Iodo-N-methylaniline ≥95%
Supplier: Matrix Scientific
074702-5G
074702-1G
102537-776EA
710.77
USD
102537-776
102537-778
4-Iodo-N-methylaniline ≥95%
4-Iodo-N-methylaniline
Hazard: Irritant
Formula:
C₇H₈IN MW: 233.0496 g/mol |
MDL Number:
MFCD04037125 CAS Number: 60577-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...