1-Pyridin-4-ylpiperidin-4-one ≥95%
Supplier: Matrix Scientific
074693-5G
074693-1G
074693-250MG
102537-740EA
2061.54
USD
102537-740
102537-736
102537-738
1-Pyridin-4-ylpiperidin-4-one ≥95%
1-Pyridin-4-ylpiperidin-4-one
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O MW: 176.2151 g/mol |
MDL Number:
MFCD09030756 CAS Number: 126832-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...