5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
Supplier: Matrix Scientific
074676-5G
074676-1G
074676-250MG
102537-650EA
2692.31
USD
102537-650
102537-648
102537-646
5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
5-Chloro-1,2,3,4-tetrahydroquinoline hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁Cl₂N MW: 204.1000 g/mol |
MDL Number:
MFCD09026769 CAS Number: 90562-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...