3,5-Dibromo-1-methylpyrazin-2(1H)-one ≥95%
Supplier: Matrix Scientific
074674-1G
074674-5G
102537-636EA
569.23
USD
102537-636
102537-638
3,5-Dibromo-1-methylpyrazin-2(1H)-one ≥95%
3,5-Dibromo-1-methylpyrazin-2(1H)-one
Hazard: Irritant
Formula:
C₅H₄Br₂N₂O MW: 267.9061 g/mol |
MDL Number:
MFCD12022617 CAS Number: 87486-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...