3-Hydroxy-4-methoxybenzonitrile ≥95%
Supplier: Matrix Scientific
074666-5G
074666-25G
102551-908EA
169.23
USD
102551-908
102537-600
3-Hydroxy-4-methoxybenzonitrile ≥95%
3-Hydroxy-4-methoxybenzonitrile
Hazard: Irritant
Formula:
C₈H₇NO₂ MW: 149.1500 g/mol |
MDL Number:
MFCD02258854 CAS Number: 52805-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...