4-Chloro-3-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

074663-5G
102537-584EA 212.75 USD
102537-584
4-Chloro-3-methylbenzonitrile ≥95%
4-Chloro-3-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₆ClN
MW: 151.5900 g/mol
MDL Number: MFCD03452793
CAS Number: 4387-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR