4-Nitrosalicylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-4-nitrobenzoic acid

074654-5G
102537-536EA 38.46 USD
102537-536
4-Nitrosalicylic acid ≥95%
4-Nitrosalicylic acid

Hazard: Irritant

Formula: C₇H₅NO₅
MW: 183.12 g/mol
MDL Number: MFCD00071505
CAS Number: 619-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR