5-Methoxyisoquinoline ≥95%

Supplier: Matrix Scientific

074652-1G 074652-5G
102537-526EA 1841.54 USD
102537-526 102537-524
5-Methoxyisoquinoline ≥95%
5-Methoxyisoquinoline

Hazard: Irritant

Formula: C₁₀H₉NO
MW: 159.1900 g/mol
MDL Number: MFCD11846297
CAS Number: 90806-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR