Ethyl-3-amino-3-methylbutanoate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-amino-3-methylbutanoate hydrochloride
074637-25G
074637-1G
074637-5G
102537-450EA
89.23
USD
102537-450
102833-702
102537-448
Ethyl-3-amino-3-methylbutanoate hydrochloride ≥95%
Ethyl-3-amino-3-methylbutanoate hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClNO₂ MW: 181.6604 g/mol |
MDL Number:
MFCD11844787 CAS Number: 85532-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...