3-Bromo-6-chloropicolinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-6-chloropyridine-2-carboxylic acid

074628-5G
102537-402EA 227.69 USD
102537-402
3-Bromo-6-chloropicolinic acid ≥95%
3-Bromo-6-chloropicolinic acid

Hazard: Irritant

Formula: C₆H₃BrClNO₂
MW: 236.4505 g/mol
MDL Number: MFCD09258777
CAS Number: 929000-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR