1-(3-(Trifluoromethyl)pyridin-2-yl)piperidin-4-one ≥95%

Supplier: Matrix Scientific

074625-1G 074625-5G
102537-386EA 533.85 USD
102537-386 102537-388
1-(3-(Trifluoromethyl)pyridin-2-yl)piperidin-4-one ≥95%
1-(3-(Trifluoromethyl)pyridin-2-yl)piperidin-4-one

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₂O
MW: 244.21 g/mol
MDL Number: MFCD09927614
CAS Number: 801306-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR