1-(4-Thiazolyl)ethanone ≥95%

Supplier: Matrix Scientific

074613-1G 074613-10G 074613-5G
102537-332EA 538.46 USD
102537-332 102537-330 102537-328
1-(4-Thiazolyl)ethanone ≥95%
1-(4-Thiazolyl)ethanone

Hazard: Irritant

Formula: C₅H₅NOS
MW: 127.1643 g/mol
MDL Number: MFCD03550493
CAS Number: 38205-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR