5-Bromo-4-(trifluoromethyl)thiazol-2-amine ≥95%

Supplier: Matrix Scientific

074598-250MG 074598-5G 074598-1G
102537-254EA 561.54 USD
102537-254 102537-252 102537-250
5-Bromo-4-(trifluoromethyl)thiazol-2-amine ≥95%
5-Bromo-4-(trifluoromethyl)thiazol-2-amine

Hazard: Irritant

Formula: C₄H₂BrF₃N₂S
MW: 247.0400 g/mol
MDL Number: MFCD09751356
CAS Number: 136411-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR