Methyl-1H-pyrrole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 1H-pyrrole-3-carboxylate
074589-25G
074589-5G
102537-206EA
452.31
USD
102537-206
102551-904
Methyl-1H-pyrrole-3-carboxylate ≥95%
Methyl-1H-pyrrole-3-carboxylate
Hazard: Irritant
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Melting Pt: 85…89 °C |
MDL Number:
MFCD00967074 CAS Number: 2703-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...