2-Chloro-6-hydroxybenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-2-hydroxybenzoic acid

074587-5G
102537-198EA 230.77 USD
102537-198
2-Chloro-6-hydroxybenzoic acid ≥95%
2-Chloro-6-hydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₅ClO₃
MW: 172.5700 g/mol
MDL Number: MFCD10687031
CAS Number: 56961-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR