Thieno[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

074586-250MG 074586-5G
102537-188EA 400 USD
102537-188 102537-192
Thieno[3,2-c]pyridine ≥95%
Thieno[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₅NS
MW: 135.1863 g/mol
MDL Number: MFCD01760493
CAS Number: 272-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR