6-Ethoxy-1H-indole ≥95%

Supplier: Matrix Scientific

074580-1G
102537-154EA 640 USD
102537-154
6-Ethoxy-1H-indole ≥95%
6-Ethoxy-1H-indole

Hazard: Irritant

Formula: C₁₀H₁₁NO
MW: 161.2000 g/mol
MDL Number: MFCD00051498
CAS Number: 37865-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR