N-(2-(1H-Indol-3-yl)ethyl)nicotinamide ≥95%

Supplier: Matrix Scientific

074574-1G
102537-126EA 1383.08 USD
102537-126
N-(2-(1H-Indol-3-yl)ethyl)nicotinamide ≥95%
N-(2-(1H-Indol-3-yl)ethyl)nicotinamide

Hazard: Irritant

Formula: C₁₆H₁₅N₃O
CAS Number: 29876-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR