5-Bromo-1H-indazol-3-ol ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-1,2-dihydro-3H-indazol-3-one
074572-250MG
074572-5G
102537-114EA
353.85
USD
102537-114
102537-118
5-Bromo-1H-indazol-3-ol ≥95%
5-Bromo-1H-indazol-3-ol
Hazard: Irritant
Formula:
C₇H₅BrN₂O MW: 213.0300 g/mol |
MDL Number:
MFCD07781638 CAS Number: 7364-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...