5-Bromo-1-methyl-1H-pyrazin-2-one ≥95%

Supplier: Matrix Scientific

074571-5G
102537-110EA 701.54 USD
102537-110
5-Bromo-1-methyl-1H-pyrazin-2-one ≥95%
5-Bromo-1-methyl-1H-pyrazin-2-one

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.0100 g/mol
MDL Number: MFCD16659841
CAS Number: 1243288-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR