Methyl-2,4-difluoro-5-nitrobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2,4-difluoro-5-nitrobenzoate
074566-10G
074566-1G
102537-082EA
107.69
USD
102537-082
102537-086
Methyl-2,4-difluoro-5-nitrobenzoate ≥95%
Methyl 2,4-difluoro-5-nitrobenzoate
Hazard: Irritant
Formula:
C₈H₅F₂NO₄ MW: 217.1264 g/mol |
MDL Number:
MFCD07779369 CAS Number: 125568-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...