(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (3R)-3-(Methylamino)pyrrolidine, 3-BOC protected

074554-1G
102537-024EA 623.08 USD
102537-024
(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥95%
(R)-tert-Butyl methyl(pyrrolidin-3-yl)carbamate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.2800 g/mol
MDL Number: MFCD09263387
CAS Number: 392338-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR