2,5,6-Trichloro-4-methylnicotinonitrile ≥95%

Supplier: Matrix Scientific

074547-250MG 074547-5G
102536-986EA 355.38 USD
102536-986 102536-990
2,5,6-Trichloro-4-methylnicotinonitrile ≥95%
2,5,6-Trichloro-4-methylnicotinonitrile

Hazard: Irritant

Formula: C₇H₃Cl₃N₂
MW: 221.47 g/mol
MDL Number: MFCD09754021
CAS Number: 63195-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR