Thieno[2,3-b]pyridin-5-amine ≥95%

Supplier: Matrix Scientific

074545-250MG
102536-978EA 1223.08 USD
102536-978
Thieno[2,3-b]pyridin-5-amine ≥95%
Thieno[2,3-b]pyridin-5-amine

Hazard: Irritant

Formula: C₇H₆N₂S
MW: 150.2 g/mol
MDL Number: MFCD14706798
CAS Number: 21344-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR