Benzyl(3-oxocyclobutyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl (3-oxocyclobutyl)carbamate3-Aminocyclobutanone, N-CBZ protected

074544-1G
102536-974EA 195.38 USD
102536-974
Benzyl(3-oxocyclobutyl)carbamate ≥95%
Benzyl(3-oxocyclobutyl)carbamate

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MW: 219.2400 g/mol
MDL Number: MFCD13152267
CAS Number: 130369-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR